5-Amino-m-anisic acid ≥97%

694428-5G 694428-1G
220090-581EA 166.92 USD
220090-581 220090-580
5-Amino-m-anisic acid ≥97%
5-Amino-m-anisic acid
Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD07783718
CAS Number: 74165-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR