(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

Synonyms: (R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (R)-(+)-BINAP, (R)-BINAP, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

693065-500MG 693065-100MG
220090-445EA 82.6 USD
220090-445 220090-444
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Formula: C₄₄H₃₂P₂
MW: 622.69 g/mol
Melting Pt: 239…242 °C
MDL Number: MFCD00010805
CAS Number: 76189-55-4
Merck Index: 13,01223

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR