(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

Synonyms: (S)-(-)-BINAP, (S)-BINAP, (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (S)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (S)-(-)-2,2'-Bis(diphenylphoshino)-1,1'-binaphthyl

693057-100MG 693057-500MG
220090-442EA 21.54 USD
220090-442 220090-443
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Formula: C₄₄H₃₂P₂
MW: 622.69 g/mol
Melting Pt: 241…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00010805
CAS Number: 76189-56-5
Merck Index: 13,01223

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR