1-Methyl-3-octylimidazolium hexafluorophosphate

69230-5G-F 69230-25G-F
220089-030EA 77.1 USD
220089-030 220090-333
1-Methyl-3-octylimidazolium hexafluorophosphate
1-Methyl-3-octylimidazolium hexafluorophosphate
Formula: C₁₂H₂₃F₆N₂P
MW: 340.29 g/mol
MDL Number: MFCD06798203
CAS Number: 304680-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR