3-Bromo-5-iodopyridine

691453-1G 691453-250MG
220090-253EA 83.46 USD
220090-253 220090-254
3-Bromo-5-iodopyridine
3-Bromo-5-iodopyridine
Formula: C₅H₃BrIN
MW: 283.89 g/mol
MDL Number: MFCD03086019
CAS Number: 233770-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR