4-Amino-3-chlorobenzoic acid ≥97%

Warning

691445-5G 691445-1G
220090-252EA 88.31 USD
220090-252 220090-251
4-Amino-3-chlorobenzoic acid ≥97%
4-Amino-3-chlorobenzoic acid
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
MDL Number: MFCD00460423
CAS Number: 2486-71-7
EINECS: 219-630-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR