Butyl cyanoacetate, Sigma-Aldrich®

Synonyms: Cyanoacetic acid butyl ester

691283-25KG 691283-1KG 691283-200KG
220089-856EA 401.54 USD
220089-856 220089-854 220089-855
Butyl cyanoacetate, Sigma-Aldrich®
Butyl cyanoacetate
Formula: C₇H₁₁NO₂
MW: 141.17 g/mol
Boiling Pt: 230.7 °C (1013 hPa)
Melting Pt: -80 °C
Density: 1.001 g/cm³ (20 °C)
MDL Number: MFCD00001941
CAS Number: 5459-58-5
EINECS: 226-730-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR