Allyl cyanoacetate ≥99.0% (GC, KF), Sigma-Aldrich®

Synonyms: Cyanoacetic acid Allyl Ester, Allyl Cyanoacetate, Allyl 2-cyanoacetate, Cyanoacetic acid allyl ester

691186-1KG 691186-200KG 691186-25KG
220089-837EA 124.37 USD
220089-837 220089-838 220089-839
Allyl cyanoacetate ≥99.0% (GC, KF), Sigma-Aldrich®
Allyl cyanoacetate
Formula: C₆H₇NO₂
MW: 125.13 g/mol
Melting Pt: –40 °C
Density: 1.07 g/cm³ (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00013817
CAS Number: 13361-32-5
EINECS: 236-423-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR