1-Decyl-3-methylimidazoliumtetrafluoroborate

Synonyms: 1-Decyl-3-methylimidazolium tetrafluoroborate

690708-5G 690708-50G
220089-064EA 104 USD
220089-064 220089-406
1-Decyl-3-methylimidazoliumtetrafluoroborate
1-Decyl-3-methylimidazoliumtetrafluoroborate
Formula: C₁₄H₂₇BF₄N₂
MW: 310.19 g/mol
MDL Number: MFCD09038853
CAS Number: 244193-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR