3-Bromo-1-butyne, Technical Grade

690643-1G 690643-5G
220090-113EA 284.31 USD
220090-113 220089-398
3-Bromo-1-butyne, Technical Grade
3-Bromo-1-butyne
Formula: C₄H₅Br
MW: 132.99 g/mol
Storage Temperature: Refrigerator
CAS Number: 18668-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR