Malononitrile, Sigma-Aldrich®

Danger

690414-10KG 690414-1KG 690414-50KG
220090-092EA 1766.15 USD
220090-092 220090-093 220090-094
Malononitrile, Sigma-Aldrich®
Malononitrile
Formula: C₃H₂N₂
MW: 66.06 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 30 - 32 °C
Density: 1.191 g/cm³ (20 °C)
Flash Pt: 112 °C
MDL Number: MFCD00001883
CAS Number: 109-77-3
EINECS: 203-703-2
UN: 2647
ADR: 6.1,II
Merck Index: 13,05733

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR