7-Bromo-2-naphthol

Synonyms: 7-Bromonaphthalen-2-ol, 7-Bromo-2-hydroxynaphthalene, 2-Bromo-7-hydroxynaphthalene

690244-5G 690244-1G
220090-078EA 163.85 USD
220090-078 220090-077
7-Bromo-2-naphthol
7-Bromo-2-naphthol
Formula: C₁₀H₇BrO
MW: 223.07 g/mol
MDL Number: MFCD00277644
CAS Number: 116230-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR