Benzyl acetoacetate, Sigma-Aldrich®

689866-25KG 689866-200KG 689866-1KG
220090-030EA 92.6 USD
220090-030 220090-029 220090-028
Benzyl acetoacetate, Sigma-Aldrich®
Benzyl acetoacetate
Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Boiling Pt: 276 °C (1013 hPa)
Density: 1.11 g/cm³ (20 °C)
MDL Number: MFCD00008785
CAS Number: 5396-89-4
EINECS: 226-416-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR