2-Bromo-4-nitrophenol

Synonyms: 1-Bromo-2-hydroxy-5-nitrobenzene

689858-1G 689858-5G
220090-026EA 107.48 USD
220090-026 220090-027
2-Bromo-4-nitrophenol
2-Bromo-4-nitrophenol
Formula: C₆H₄BrNO₃
MW: 218.01 g/mol
MDL Number: MFCD06656567
CAS Number: 5847-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR