2-Amino-4-bromophenol

Synonyms: 5-Bromo-2-hydroxyaniline

689629-5G 689629-1G
220087-862EA 221.54 USD
220087-862 220087-861
2-Amino-4-bromophenol
2-Amino-4-bromophenol
Formula: C₆H₆BrNO
MW: 188.02 g/mol
MDL Number: MFCD00235171
CAS Number: 40925-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR