2-Bromo-4-nitropyridine

689610-5G 689610-1G
220087-859EA 255.23 USD
220087-859 220087-858
2-Bromo-4-nitropyridine
2-Bromo-4-nitropyridine
Formula: C₅H₃BrN₂O₂
MW: 203 g/mol
MDL Number: MFCD03701383
CAS Number: 6945-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR