1-Ethyl-3-methylimidazolium acetate

Synonyms: 1-Ethyl-3-methylimidazolium Acetate

689483-5G 689483-50G
220089-931EA 185 USD
220089-931 220089-930
1-Ethyl-3-methylimidazolium acetate
1-Ethyl-3-methylimidazolium acetate
Formula: C₈H₁₄N₂O₂
MW: 170.21 g/mol
MDL Number: MFCD06798186
CAS Number: 143314-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR