Sodium dehydracetate monohydrate 98.0-101.0% (w/w), Sigma-Aldrich®

Warning

Synonyms: Sodium 3-Acetyl-4-hydroxy-6-methyl-2-oxo-4(2H)-pyranolate, Sodium Dehydroacetate monohydrate, Sodium dehydroacetate monohydrate, Dehydroacetic acid sodium salt monohydrate, Sodium 1-(3,4-dihydro-6-methyl-2,4-dioxo-2H-pyran-3-ylidene)ethonolate monohydrate, 3-Acetyl-4-hydroxy-6-methyl-2-pyrone sodium salt, Sodium Dehydroacetate Monohydrate, Dehydracetic acid sodium salt monohydrate

689424-1KG 689424-10KG 689424-40KG
220089-922EA 98.65 USD
220089-922 220089-921 220089-923
Sodium dehydracetate monohydrate 98.0-101.0% (w/w), Sigma-Aldrich®
Sodium dehydracetate monohydrate
Formula: C₈H₇NaO₄·1H₂O
MW: 208.15 g/mol
MDL Number: MFCD00040583
CAS Number: 64039-28-7
EINECS: 224-580-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR