1-Butyl-3-methylimidazoliumtrifluoroacetate

Synonyms: 1-Butyl-3-methylimidazolium Trifluoroacetate

689270-50G 689270-5G
220089-906EA 1129.62 USD
220089-906 220089-907
1-Butyl-3-methylimidazoliumtrifluoroacetate
1-Butyl-3-methylimidazoliumtrifluoroacetate
Formula: C₁₀H₁₅F₃N₂O₂
MW: 252.24 g/mol
MDL Number: MFCD08458921
CAS Number: 174899-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR