3,4-Dihydroxy-3-cyclobutene-1,2-dione, Sigma-Aldrich®

Synonyms: Squaric acid, 1,2-Dihydroxy-3,4-cyclobutenedione, Quadratic acid, 1,2-Dihydroxy-1-cyclobutene-3,4-dione, Diketocyclobutenediol

689203-2.5KG 689203-100G 689203-50KG
220089-894EA 131.54 USD
220089-894 220089-893 220089-895
3,4-Dihydroxy-3-cyclobutene-1,2-dione, Sigma-Aldrich®
3,4-Dihydroxy-3-cyclobutene-1,2-dione
Formula: C₄H₂O₄
MW: 114.06 g/mol
Melting Pt: 293 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00001334
CAS Number: 2892-51-5
EINECS: 220-761-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR