Ethyl-4,4,4-trifluoroacetoacetate, Sigma-Aldrich®

Synonyms: Ethyl trifluoroacetoacetate, Ethyl 4,4,4-trifluoro-3-oxobutanoate

689106-25KG 689106-5KG
220089-816EA 2411.54 USD
220089-816 220089-817
Ethyl-4,4,4-trifluoroacetoacetate, Sigma-Aldrich®
Ethyl-4,4,4-trifluoroacetoacetate
Formula: C₆H₇F₃O₃
MW: 184.11 g/mol
Boiling Pt: 131 °C (1013 hPa)
Melting Pt: -39 °C
Density: 1.2586 g/cm³ (15.5 °C)
MDL Number: MFCD00000424
CAS Number: 372-31-6
EINECS: 206-750-7
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR