Ethyl acetoacetate, Sigma-Aldrich®

Warning

Synonyms: Acetoacetic acid ethyl ester, EAA

688983-1KG 688983-240KG 688983-25KG
220089-002EA 132.46 USD
220089-002 220089-003 220089-004
Ethyl acetoacetate, Sigma-Aldrich®
Ethyl acetoacetate
Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 180.8 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.0188 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00009199
CAS Number: 141-97-9
EINECS: 205-516-1
Merck Index: 13,03793

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR