Methyl acetoacetate, Sigma-Aldrich®

Warning

Synonyms: Methyl 3-oxobutanoate

688886-1KG 688886-25KG 688886-250KG
220089-805EA 66.77 USD
220089-805 220089-389 220089-388
Methyl acetoacetate, Sigma-Aldrich®
Methyl acetoacetate
Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 170 °C (1013 hPa)
Melting Pt: -39 °C
Density: 1.076 g/cm³ (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD00008784
CAS Number: 105-45-3
EINECS: 203-299-8
Merck Index: 12,06090

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR