tert-Butyl acetoacetate, Sigma-Aldrich®

Synonyms: tert-Butyl Acetoacetate

688770-230KG 688770-25KG 688770-1KG
220089-794EA 175.38 USD
220089-794 220089-795 220089-793
tert-Butyl acetoacetate, Sigma-Aldrich®
tert-Butyl acetoacetate
Formula: C₈H₁₄O₃
MW: 158.2 g/mol
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: -38 °C
Density: 0.9756 g/cm³ (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00008811
CAS Number: 1694-31-1
EINECS: 216-904-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR