3-Amino-2-(trifluoromethyl)pyridine

Synonyms: 2-(Trifluoromethyl)-3-pyridylamine, 2-(Trifluoromethyl)pyridin-3-amine

686972-250MG 686972-1G
220089-640EA 156.15 USD
220089-640 220089-639
3-Amino-2-(trifluoromethyl)pyridine
3-Amino-2-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MW: 162.11 g/mol
MDL Number: MFCD09753488
CAS Number: 106877-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR