3-Amino-4-bromobenzoic acid ≥97%

Synonyms: 3-Amino-4-bromobenzoic Acid

686433-5G 686433-25G
220089-612EA 239.08 USD
220089-612 220089-611
3-Amino-4-bromobenzoic acid ≥97%
3-Amino-4-bromobenzoic acid
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
MDL Number: MFCD00579106
CAS Number: 2840-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR