6-Bromo-m-toluidine ≥97%

686409-5G 686409-25G
220089-607EA 42.54 USD
220089-607 220089-606
6-Bromo-m-toluidine ≥97%
6-Bromo-m-toluidine
Formula: C₇H₈BrN
MW: 186.05 g/mol
MDL Number: MFCD01806278
CAS Number: 53078-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR