(R)-(+)-4-Bromo-α-methylbenzyl alcohol ≥95%

Synonyms: (R)-1-(4-Bromophenyl)ethanol

686336-1G 686336-250MG
220089-596EA 179.85 USD
220089-596 220089-597
(R)-(+)-4-Bromo-α-methylbenzyl alcohol ≥95%
(R)-(+)-4-Bromo-α-methylbenzyl alcohol
Formula: C₈H₉BrO
MW: 201.06 g/mol
MDL Number: MFCD06201861
CAS Number: 76155-78-7

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR