(S)-(-)-1-Phenylethanol ≥97%

Warning

Synonyms: (S)-(-)-α-Methylbenzyl alcohol, (S)-(-)-sec-Phenethyl alcohol, (S)-1-Phenylethanol

685836-25G 685836-5G
220089-538EA 663.38 USD
220089-538 220089-539
(S)-(-)-1-Phenylethanol ≥97%
(S)-(-)-1-Phenylethanol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 207 °C (1013 hPa)
Density: 1.018 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064264
CAS Number: 1445-91-6
UN: 2937
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR