(R)-(+)-1-Phenylethanol ≥97%

Warning

Synonyms: (R)-1-Phenylethanol, (R)-(+)-α-Methylbenzyl alcohol, (R)-(+)-sec-Phenethyl alcohol

685828-25G 685828-5G
220089-536EA 147.54 USD
220089-536 220089-537
(R)-(+)-1-Phenylethanol ≥97%
(R)-(+)-1-Phenylethanol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 207 °C (1013 hPa)
Density: 1.012 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064263
CAS Number: 1517-69-7
UN: 2937
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR