2-Amino-5-(trifluoromethyl)pyridine

Synonyms: 5-(Trifluoromethyl)pyridin-2-amine, 5-(Trifluoromethyl)-2-pyridylamine

684716-1G 684716-250MG
220088-993EA 63.22 USD
220088-993 220089-431
2-Amino-5-(trifluoromethyl)pyridine
2-Amino-5-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MW: 162.11 g/mol
MDL Number: MFCD00042164
CAS Number: 74784-70-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR