8-Chloroquinaldine ≥97%

Synonyms: 8-Chloro-2-methylquinoline

683914-5G 683914-1G
220089-322EA 109.85 USD
220089-322 220089-321
8-Chloroquinaldine ≥97%
8-Chloroquinaldine
Formula: C₁₀H₈ClN
MW: 177.63 g/mol
MDL Number: MFCD00051764
CAS Number: 3033-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR