2-Cyanophenylboronic acid pinacol ester ≥97%

Synonyms: 2-Cyanobenzeneboronic acid pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 2-Cyanophenylboronic acid, pinacol ester

683884-1G 683884-5G
220087-844EA 42.23 USD
220087-844 220087-845
2-Cyanophenylboronic acid pinacol ester ≥97%
2-Cyanophenylboronic acid pinacol ester
Formula: C₁₃H₁₆BNO₂
MW: 229.09 g/mol
MDL Number: MFCD04038747
CAS Number: 214360-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR