2-(Trifluoromethoxy)phenylboronic acid

Synonyms: 2-Trifluoromethoxyphenylboronic acid, 2-(Trifluoromethoxy)benzeneboronic acid, (2-(Trifluoromethoxy)phenyl)metaboric acid, Dihydroxy(2-(trifluoromethoxy)phenyl)borane

683787-1G 683787-5G
220089-308EA 23.46 USD
220089-308 220089-309
2-(Trifluoromethoxy)phenylboronic acid
2-(Trifluoromethoxy)phenylboronic acid
Formula: C₇H₆BF₃O₃
MW: 205.93 g/mol
MDL Number: MFCD01320763
CAS Number: 175676-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR