1-Chloro-4-ethylbenzene ≥97%

Synonyms: 4-Chloro(ethylbenzene)

683744-25G 683744-5G
220089-301EA 438.46 USD
220089-301 220089-302
1-Chloro-4-ethylbenzene ≥97%
1-Chloro-4-ethylbenzene
Formula: C₈H₉Cl
MW: 140.61 g/mol
MDL Number: MFCD00040855
CAS Number: 622-98-0
EINECS: 210-763-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR