Methyl-3-amino-4-methoxybenzoate ≥97%

Synonyms: Methyl 3-amino-4-methoxybenzoate, 3-Amino-4-methoxybenzoic acid methyl ester

683736-25G 683736-5G
220089-299EA 171.23 USD
220089-299 220089-300
Methyl-3-amino-4-methoxybenzoate ≥97%
Methyl-3-amino-4-methoxybenzoate
Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
MDL Number: MFCD00017196
CAS Number: 24812-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR