6-Methyl-5-nitroquinoline

683485-1G 683485-5G
220089-270EA 126 USD
220089-270 220089-271
6-Methyl-5-nitroquinoline
6-Methyl-5-nitroquinoline
Formula: C₁₀H₈N₂O₂
MW: 188.19 g/mol
MDL Number: MFCD00024021
CAS Number: 23141-61-9
EINECS: 245-447-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR