Palladium(II) acetylacetonate

Danger

Synonyms: Palladium (II) 2,4-pentanedionate, Bis(2,4-pentanedionato-O,O')palladium (II), Palladium (II) bis(2,4-pentanedionato-O,O'), Palladium(II) 2,4-pentanedionate

683388-500MG 683388-2G
220089-256EA 34.75 USD
220089-256 220089-255
Palladium(II) acetylacetonate
Palladium(II) acetylacetonate
Formula: C₁₀H₁₄O₄Pd
MW: 304.64 g/mol
Boiling Pt: ? 190 °C (1013 hPa)
Melting Pt: 190 °C
MDL Number: MFCD00000025
CAS Number: 14024-61-4
EINECS: 237-859-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR