Palladium(II) acetate ≥98%

Synonyms: Palladium diacetate, Palladium (II) acetate

683124-500MG 683124-5G
220087-547EA 62.78 USD
220087-547 220087-548
Palladium(II) acetate ≥98%
Palladium(II) acetate
Formula: C₄H₆O₄Pd
MW: 224.51 g/mol
Melting Pt: 205 °C
MDL Number: MFCD00012453
CAS Number: 3375-31-3
EINECS: 222-164-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR