2-Bromobenzophenone

Synonyms: (2-Bromophenyl)(phenyl)methanone

682926-5G 682926-1G
220088-972EA 157.23 USD
220088-972 220088-971
2-Bromobenzophenone
2-Bromobenzophenone
Formula: C₁₃H₉BrO
MW: 261.12 g/mol
Boiling Pt: 345 °C (1013 hPa)
Melting Pt: 42 °C
Density: 1.4309 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00032462
CAS Number: 13047-06-8
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR