2,4,6-Trimethylbenzoyl chloride ≥97%

Synonyms: 2,4,6-Trimethylbenzene-1-carbonyl chloride, 2,4,6-Trimethylbenzoyl Chloride

682519-5G 682519-1G
220088-012EA 289.69 USD
220088-012 220088-011
2,4,6-Trimethylbenzoyl chloride ≥97%
2,4,6-Trimethylbenzoyl chloride
Formula: C₁₀H₁₁ClO
MW: 182.65 g/mol
Boiling Pt: 48 °C (0.05 -rr)
Density: 1.0967 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00013650
CAS Number: 938-18-1
EINECS: 213-339-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR