3-Chlorobenzoylacetonitrile ≥97%

Synonyms: 3'-Chloro-2-cyanoacetophenone, (3-Chlorobenzoyl)acetonitrile, 3-(3-Chlorophenyl)-3-oxopropanenitrile

681857-5G 681857-1G
220089-132EA 76.46 USD
220089-132 220089-131
3-Chlorobenzoylacetonitrile ≥97%
3-Chlorobenzoylacetonitrile
Formula: C₉H₆ClNO
MW: 179.61 g/mol
MDL Number: MFCD00067891
CAS Number: 21667-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR