4-Fluorobenzoylacetonitrile ≥97%

Synonyms: 3-(4-Fluorophenyl)-3-oxopropanenitrile

681822-1G 681822-5G
220089-126EA 45.77 USD
220089-126 220089-127
4-Fluorobenzoylacetonitrile ≥97%
4-Fluorobenzoylacetonitrile
Formula: C₉H₆FNO
MW: 163.15 g/mol
MDL Number: MFCD00662062
CAS Number: 4640-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR