3-(Benzyloxy)benzoic acid ≥97%

Synonyms: 3-Benzyloxybenzoic acid

681717-5G 681717-1G
220089-113EA 101.23 USD
220089-113 220089-112
3-(Benzyloxy)benzoic acid ≥97%
3-(Benzyloxy)benzoic acid
Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
MDL Number: MFCD00527901
CAS Number: 69026-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR