Ethyl indole-3-carboxylate

Synonyms: Indole-3-carboxylic acid ethyl ester, Ethyl 1H-indole-3-carboxylate

681466-1G 681466-5G
220089-086EA 438.46 USD
220089-086 220089-087
Ethyl indole-3-carboxylate
Ethyl indole-3-carboxylate
Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
MDL Number: MFCD00228454
CAS Number: 776-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR