Potassium(bromomethyl)trifluoroborate ≥90%

Synonyms: (Bromomethyl)trifluoroboric acid potassium salt, Potassium (bromomethyl)trifluoroborate

681342-5G 681342-1G
220087-033EA 32.54 USD
220087-033 220087-032
Potassium(bromomethyl)trifluoroborate ≥90%
Potassium(bromomethyl)trifluoroborate
Formula: CH₂BBrF₃K
MW: 200.83 g/mol
MDL Number: MFCD09265154
CAS Number: 888711-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR