1-Bromo-3-fluoro-4-nitrobenzene

Synonyms: 4-Bromo-2-fluoronitrobenzene

680931-500MG 680931-5G
220088-918EA 438.46 USD
220088-918 220088-919
1-Bromo-3-fluoro-4-nitrobenzene
1-Bromo-3-fluoro-4-nitrobenzene
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
MDL Number: MFCD01930221
CAS Number: 321-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR