5-Methoxy-m-cresol

Warning

Synonyms: 3-Hydroxy-5-methylanisol, 1-Hydroxy-3-methoxy-5-methylbenzene, 3-Methoxy-5-methylphenol, 3-Hydroxy-5-methoxytoluene

680893-1G 680893-5G
220088-912EA 438.46 USD
220088-912 220088-913
5-Methoxy-m-cresol
5-Methoxy-m-cresol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
MDL Number: MFCD00059261
CAS Number: 3209-13-0
EINECS: 221-716-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR