1-Boc-5-methoxyindole

Synonyms: tert-Butyl 5-methoxy-1H-indole-1-carboxylate, N-Boc-5-methoxyindole

680613-10G 680613-1G
220088-878EA 393.08 USD
220088-878 220088-879
1-Boc-5-methoxyindole
1-Boc-5-methoxyindole
Formula: C₁₄H₁₇NO₃
MW: 247.29 g/mol
MDL Number: MFCD08272248
CAS Number: 99275-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR