2-Chloro-5-(trifluoromethyl)phenylboronic acid

Synonyms: 2-Chloro-5-(trifluoromethyl)phenylboronic Acid, Dihydroxy(2-chloro-5-(trifluoromethyl)phenyl)borane, (2-Chloro-5-(trifluoromethyl)phenyl)metaboric acid, 2-Chloro-5-(trifluoromethyl)benzeneboronic acid, 2-Chloro-5-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride)

680524-1G 680524-10G
220088-861EA 438.46 USD
220088-861 220088-860
2-Chloro-5-(trifluoromethyl)phenylboronic acid
2-Chloro-5-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BClF₃O₂
MW: 224.37 g/mol
MDL Number: MFCD00797335
CAS Number: 182344-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR